Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(bis(2-(tert-butoxy)-2-oxoethyl)amino)acetic acid | 171557-31-6 | 97.0% | 303.35 g/mol | C14H25NO6 | 5 G
SDP

Catalog No. 5000209347
Encompass_Preferred

2-(bis(2-(tert-butoxy)-2-oxoethyl)amino)acetic acid is a tert-butoxy-protected glycine derivative supplied as a solid reagent for research and analytical applications. It is commonly used as an intermediate in peptide synthesis and organic chemistry.

  • Chemical formula C14H25NO6.
  • Molecular weight 303.35 g/mol.
  • Purity 97.0%.
  • Physical form solid (powder).
  • Available sizes 1 g, 5 g, 10 g, 50 g (quote).
  • Storage powder: -20 °C (3 years) or 4 °C (2 years); in solvent: -80 °C (6 months) or -20 °C (1 month).

Catalog No. 50-002-09347 Supplier Medchemexpress LLC Supplier No. HYW0406865G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(bis(2-(tert-butoxy)-2-oxoethyl)amino)acetic acid is a tert-butoxy-protected glycine derivative supplied as a solid reagent for research and analytical applications. It is commonly used as an intermediate in peptide synthesis and organic chemistry.

  • Chemical formula C14H25NO6.
  • Molecular weight 303.35 g/mol.
  • Purity 97.0%.
  • Physical form solid (powder).
  • Available sizes 1 g, 5 g, 10 g, 50 g (quote).
  • Storage powder: -20 °C (3 years) or 4 °C (2 years); in solvent: -80 °C (6 months) or -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.