Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[Bis(methylseleno)]propane, Thermo Scientific™

Catalog No. AC123920050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC123920050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC123920050 Supplier Thermo Scientific Chemicals Supplier No. 123920050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56051-06-0
Molecular Formula C5H12Se2
Molecular Weight (g/mol) 230.093
InChI Key KPGCXISRRPRULE-UHFFFAOYSA-N
Synonym 2,2-bis methylselanyl propane, bis methylseleno propane, 2-bis methylseleno propane
PubChem CID 2723678
IUPAC Name 2,2-bis(methylselanyl)propane
SMILES CC(C)([Se]C)[Se]C

Specifications

Linear Formula (CH3)2C(SeCH3)2
Quantity 5 g
Formula Weight 230.07
Chemical Name or Material 2-[Bis(methylseleno)]propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.