Learn More
USA Scientific Inc 2 blk position dry heat bath
Catalog No. 502744885
Encompass
Thermal-Lok two block position dry heat bath with two 1.5/2.0 mL blocks, digital display, 5C above ambient to 150C, 1-999 minute timer
Catalog No. 50-274-4885 Supplier USA Scientific Inc Supplier No. 25301102
Thermal-Lok two block position dry heat bath with two 1.5/2.0 mL blocks, digital display, 5C above ambient to 150C, 1-999 minute timer