Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Boc-Amino)Ethanethiol

Catalog No. 45891025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-891-025ML 25 mL
45-891-05ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 45-891-025ML Supplier Sigma Aldrich Supplier No. 45891025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Boc-Amino)Ethanethiol

Specifications

CAS 67385-09-5
Density 1.049 g/mL at 20°C
Boiling Point 68°C (0.3 mmHg, lit.)
Molecular Formula C7H15NO2S
Refractive Index n20/D 1.474
Linear Formula HSCH2CH2NHCO2C(CH3)3
MDL Number MFCD00274335
Quantity 25 mL
Synonym tert-Butyl N-(2-mercaptoethyl)carbamate
Molecular Weight (g/mol) 177.26 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.