Learn More
Selleck Chemical LLC 2-BP-E0120-10MM/1ML
SDP
Catalog No. 503135679
Encompass_Preferred
2-BP is a nonmetabolisable analogue of palmitate and acts as a palmitoylation inhibitor which inhibits multiple palmitoyl-acyl transferase (PAT) activity towards MyrGCK(NBD) and FarnCNRas(NBD) with identical IC50s of approx 4 M 2-Bromohexadecanoic acid can directly and irreversibly inhibit the palmitoyltransferase activity of all DHHC (Asp-His-His-Cys) proteins
Catalog No. 50-313-5679 Supplier Selleck Chemical LLC Supplier No. E012010MM/1ML
Add to Cart
Add to Cart
2-BP is a nonmetabolisable analogue of palmitate and acts as a palmitoylation inhibitor which inhibits multiple palmitoyl-acyl transferase (PAT) activity towards MyrGCK(NBD) and FarnCNRas(NBD) with identical IC50s of approx 4 M 2-Bromohexadecanoic acid can directly and irreversibly inhibit the palmitoyltransferase activity of all DHHC (Asp-His-His-Cys) proteins