Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromo-1,1-dimethoxyethane

Catalog No. 24250025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-250-025G 25 g
24-250-0100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-250-025G Supplier Sigma Aldrich Supplier No. 24250025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Bromo-1,1-dimethoxyethane (Bromoacetaldehyde dimethyl acetal) was used in the synthesis of 2,3-O-acetal via reaction with 2,3-diol in the presence of 10-camphorsulphonic acid (CSA).

Specifications

CAS 7252-83-7
Density 1.43 g/mL (at 25°C (literature))
Boiling Point 148°C to 150°C (lit.)
Molecular Formula C4H9BrO2
Refractive Index n20/D 1.445 (literature)
Linear Formula BrCH2CH(OCH3)2
MDL Number MFCD00000213
Quantity 25 g
Synonym Bromoacetaldehyde dimethyl acetal
Molecular Weight (g/mol) 169.02
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.