Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(1-methyl-1h-benzimidazol-2-yl)-1-ethanone, 97%, Thermo Scientific™

Catalog No. CC03805DE
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03805DE 5 g Amber Glass Bottle
CC03805DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03805DE Supplier Thermo Scientific Chemicals Supplier No. CC03805DE

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 56653-43-1
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.10
MDL Number MFCD06200854
InChI Key KHFRWYRANOMZCP-UHFFFAOYSA-N
Synonym 2-bromo-1-1-methyl-1h-benzimidazol-2-yl-1-ethanone, 2-bromo-1-1-methyl-1,3-benzodiazol-2-yl ethanone, 2-bromo-1-1-methyl-1h-benzo d imidazol-2-yl-1-ethanone, 1-methyl-2-bromoacetyl-1h-benzimidazole, 2-bromo-1-1-methylbenzimidazol-2-yl ethanone, 2-bromo-1-1-methyl-1h-benzimidazol-2-yl ethanone, 2-bromo-1-1-methylbenzimidazol-2-yl ethan-1-one, 2-bromo-1-1-methyl-1h-benzoimidazol-2-yl-ethanone, 2-bromo-1-1-methyl-1h-benzoimidazol-2-yl ethanone, 2-bromo-1-1-methyl-1h-benzo d imidazol-2-yl ethanone
PubChem CID 2795119
IUPAC Name 2-bromo-1-(1-methylbenzimidazol-2-yl)ethanone
SMILES CN1C(=NC2=CC=CC=C12)C(=O)CBr

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(1-methyl-1H-benzimidazol-2-yl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.