Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one, Thermo Scientific™

Catalog No. CC01305DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC01305DE 5 g
CC01305CB 250 mg
CC01305DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC01305DE Supplier Thermo Scientific Chemicals Supplier No. CC01305DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4629-54-3
Molecular Formula C10H9BrO3
Molecular Weight (g/mol) 257.08
MDL Number MFCD00099402
InChI Key CSSHRKYOZTZFCX-UHFFFAOYSA-N
Synonym 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 2-bromo-1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4 dioxin-6-yl ethanone, 2-bromo-1-2,3-dihydrobenzo 1,4-dioxin-6-yl ethanone, 1,4-benzodioxan-6-ylbromomethyl ketone, 2-bromo-1-2,3-dihydro-benzo 1,4 dioxin-6-yl-ethanone, ethanone,2-bromo-1-2,3-dihydro-1,4-benzodioxin-6-yl
PubChem CID 2776171
SMILES BrCC(=O)C1=CC=C2OCCOC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name 2-bromo-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
Formula Weight 257.08
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(2, 3-dihydro-1, 4-benzodioxin-6-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.