Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-bromo-1-(2-phenoxyphenyl)ethanone, Thermo Scientific™

Catalog No. CC49905DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC49905DA 1 g Amber Glass Bottle
CC49905CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC49905DA Supplier Thermo Scientific Chemicals Supplier No. CC49905DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94402-42-3
Molecular Formula C14H11BrO2
Molecular Weight (g/mol) 291.144
MDL Number MFCD08435906
InChI Key UWRDQDVDGPRCKR-UHFFFAOYSA-N
Synonym 2-bromo-1-2-phenoxyphenyl ethanone, 2-bromo-1-2-phenoxyphenyl ethan-1-one, 2-phenoxy phenacyl bromide, 2-bromoacetyl diphenyl ether
PubChem CID 13074046
IUPAC Name 2-bromo-1-(2-phenoxyphenyl)ethanone
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 291.14
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(2-phenoxyphenyl)ethanone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.