Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-[3,5-di(trifluoromethyl)phenyl]ethan-1-one, 95%, Thermo Scientific™

Catalog No. SPB07614EB
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB07614EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB07614EB Supplier Thermo Scientific Chemicals Supplier No. SPB07614EB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131805-94-2
Molecular Formula C10H5BrF6O
Molecular Weight (g/mol) 335.043
MDL Number MFCD00792434
InChI Key ZEKBFXJTIAEUOF-UHFFFAOYSA-N
Synonym 1-3,5-bis trifluoromethyl phenyl-2-bromoethanone, 3',5'-bis trifluoromethyl-2-bromoacetophenone, 1-3,5-bis trifluoromethyl phenyl-2-bromoethan-1-one, 3,5-bistrifluoromethyl-phenacyl bromide, 3,5-bis trifluoromethyl phenacyl bromide, ethanone,1-3,5-bis trifluoromethyl phenyl-2-bromo, 3,5-bis trifluoromethyl phenacylbromide, a-bromo-3',5'-bis trifluoromethyl acetophenone, 2-bromo-1-3,5-bis trifluoromethyl phenyl ethanone
PubChem CID 2736170
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]-2-bromoethanone
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)CBr

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Linear Formula (CF3)2C6H3C(O)CH2Br
Quantity 25 g
Formula Weight 335.04
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-[3, 5-di(trifluoromethyl)phenyl]ethan-1-one
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.