Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,3,5-triisopropylbenzene 95.0+%, TCI America™
SDP

Catalog No. B351325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B351325G 25 g
B35135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B351325G Supplier TCI America Supplier No. B351325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21524-34-5
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.253
MDL Number MFCD00051547
InChI Key FUMMYHVKFAHQST-UHFFFAOYSA-N
Synonym 2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf
PubChem CID 140846
IUPAC Name 2-bromo-1,3,5-tri(propan-2-yl)benzene
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C

Specifications

Color Yellow
Boiling Point 148°C
Quantity 25 g
Formula Weight 283.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-1,3,5-triisopropylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.