Learn More
Medchemexpress LLC 2-bromo-1,3-cyclohexanedione | 60060-44-8 | MFCD00971951 | 99.8% | 191.02 g/mol | C6H7BrO2 | 1 G
SDP
Catalog No. 5000301271
Encompass_Preferred
2-Bromo-1,3-cyclohexanedione is a brominated cyclohexanedione used as a drug intermediate in organic synthesis. It is supplied as a high-purity solid and serves as a versatile building block for the preparation of heterocycles and other active pharmaceutical compounds.
  • High purity (≈99.8%) suitable for research synthesis.
  • Solid form allows convenient weighing and storage.
  • Molecular formula C6H7BrO2, molecular weight 191.02 g/mol.
  • Useful intermediate for heterocycle and active pharmaceutical ingredient synthesis.
  • Available in research-scale pack sizes for small-batch work.
Catalog No. 50-003-01271 Supplier Medchemexpress LLC Supplier No. HYW0276901G
2-Bromo-1,3-cyclohexanedione is a brominated cyclohexanedione used as a drug intermediate in organic synthesis. It is supplied as a high-purity solid and serves as a versatile building block for the preparation of heterocycles and other active pharmaceutical compounds.
  • High purity (≈99.8%) suitable for research synthesis.
  • Solid form allows convenient weighing and storage.
  • Molecular formula C6H7BrO2, molecular weight 191.02 g/mol.
  • Useful intermediate for heterocycle and active pharmaceutical ingredient synthesis.
  • Available in research-scale pack sizes for small-batch work.