Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,3-difluoro-5-iodobenzene 98.0+%, TCI America™
SDP

Catalog No. B30735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30735G 5 g
B307325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30735G Supplier TCI America Supplier No. B30735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 155906-10-8
Molecular Formula C6H2BrF2I
Molecular Weight (g/mol) 318.89
MDL Number MFCD06797752
InChI Key WOLGNRYEQPISPB-UHFFFAOYSA-N
PubChem CID 2761406
IUPAC Name 2-bromo-1,3-difluoro-5-iodobenzene
SMILES FC1=CC(I)=CC(F)=C1Br

Specifications

Melting Point 66°C
Quantity 5 g
Formula Weight 318.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1,3-difluoro-5-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.