Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,3-thiazole-4-carboxylic acid, 98%

Catalog No. AC454600010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454600010 1 g
AC454600050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454600010 Supplier Thermo Scientific Chemicals Supplier No. 454600010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5198-88-9 | C4H2BrNO2S | 208.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5198-88-9
Molecular Formula C4H2BrNO2S
Molecular Weight (g/mol) 208.03
InChI Key BEGREHRAUWCAHV-UHFFFAOYSA-N
Synonym 2-bromo-4-thiazolecarboxylic acid, 2-bromothiazole-4-carboxylic acid, 2-bromo-thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 2-bromo, 2-bromo-4-thiazolecarboxylicacid, 2-bromothiazole-4-carboxylicacid, 2-bromo-4-carboxy-1,3-thiazole, pubchem22900, acmc-1avkr, 2-bromo-4-carboxythiazole
PubChem CID 2763209
IUPAC Name 2-bromo-1,3-thiazole-4-carboxylic acid
SMILES C1=C(N=C(S1)Br)C(=O)O

Specifications

Melting Point 204.0°C to 205.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 208.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-1,3-thiazole-4-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.