Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,4-benzoquinone 93.0+%, TCI America™
SDP

Catalog No. B16015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16015G 5 g
B16011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16015G Supplier TCI America Supplier No. B16015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3958-82-5
Molecular Formula C6H3BrO2
Molecular Weight (g/mol) 186.992
MDL Number MFCD00189377
InChI Key PYEQXZOIQJWYAH-UHFFFAOYSA-N
Synonym 2-Bromocyclohexadiene-1,4-dione, Bromoquinone
PubChem CID 151181
IUPAC Name 2-bromocyclohexa-2,5-diene-1,4-dione
SMILES C1=CC(=O)C(=CC1=O)Br

Specifications

Melting Point 55°C
Quantity 5 g
Formula Weight 186.99
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1,4-benzoquinone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.