Learn More
eMolecules​ 2632-13-5 | 2-Bromo-1-(4-methoxyphenyl)ethanone | Oakwood Chemical | MFCD00000201 | 229.073 | C9H9BrO2 | 97.000 | COc1ccc(cc1)C(=O)CBr | 5g | 537676305
Catalog No. NC2228499 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Bromo-1-(4-methoxyphenyl)ethanone | Oakwood Chemical | 2632-13-5 | MFCD00000201 | 229.073 | C9H9BrO2 | 97.000 | COc1ccc(cc1)C(=O)CBr | 5g | 537676305
This item is not returnable. View return policy
Catalog No. NC2228499 Supplier eMolecules​ Supplier No. 537676305
2-Bromo-1-(4-methoxyphenyl)ethanone | Oakwood Chemical | 2632-13-5 | MFCD00000201 | 229.073 | C9H9BrO2 | 97.000 | COc1ccc(cc1)C(=O)CBr | 5g | 537676305