Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(4-morpholinophenyl)-1-ethanone, 97%, Thermo Scientific™

Catalog No. CC17405DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC17405DA 1 g
CC17405CB 250 mg
CC17405DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC17405DA Supplier Thermo Scientific Chemicals Supplier No. CC17405DA
Add to Cart

Chemical Identifiers

CAS 210832-85-2
Molecular Formula C12H14BrNO2
Molecular Weight (g/mol) 284.15
MDL Number MFCD03783555
InChI Key OUGMZFJPRSTGMJ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-morpholinophenyl-1-ethanone, 2-bromo-1-4-morpholinophenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethanone, 2-bromo-1-4-4-morpholinyl phenyl ethanone, 2-bromo-1-4-morpholin-4-yl-phenyl ethanone, 2-bromo-1-4-morpholin-4-yl phenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one, pubchem23338, acmc-1cqxm, alpha-bromo-4'-morpholinoacetophenone
PubChem CID 2795357
SMILES BrCC(=O)C1=CC=C(C=C1)N1CCOCC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-bromo-1-[4-(morpholin-4-yl)phenyl]ethan-1-one
Formula Weight 284.15
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-morpholinophenyl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.