Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-chloro-2-methylpropane, 90+%

Catalog No. AAA1997414
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1997414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1997414 Supplier Thermo Scientific Chemicals Supplier No. A1997414
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2074-80-8 | C4H8BrCl | 171.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2074-80-8
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.462
MDL Number MFCD00049143
InChI Key YEZMNTBSGDUYBT-UHFFFAOYSA-N
Synonym 2-bromo-1-chloro-2-methyl-propane, acmc-20ao39, 2-brom-2-methyl-3-chlorpropan, 2-bromo-1-chloro-2-methylpropane, 90+%
PubChem CID 137422
IUPAC Name 2-bromo-1-chloro-2-methylpropane
SMILES CC(C)(CCl)Br

Specifications

Density 1.44
Boiling Point 127°C to 128°C
Flash Point 37°C (98°F)
Odor Aromatic
Refractive Index 1.474
Quantity 25 g
UN Number UN1993
Formula Weight 171.47
Percent Purity ≥90%
Chemical Name or Material 2-Bromo-1-chloro-2-methylpropane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.