Learn More
2-Bromo-1-chloropropane Solution, 20,000 μg/mL in Methanol, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Packaging | Ampule |
| CAS | 3017-95-6 |
| Linear Formula | C3H6 BrCl |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Formula Weight | 157.44 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 20,000 μg/mL in methanol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.