Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-indanol, 99%, Thermo Scientific™

Catalog No. AC107052500
Change view
Click to view available options
Quantity:
10 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC107052500 250 g
AC107050100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107052500 Supplier Thermo Scientific Chemicals Supplier No. 107052500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5400-80-6
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00003798
InChI Key RTESDSDXFLYAKZ-UHFFFAOYNA-N
Synonym 2-bromo-1-indanol, 2-bromoindan-1-ol, 1-hydroxy-2-bromoindan, 2-bromo-1-hydroxyindane, 2-bromo-1-hydroxyhydrindene, 1h-inden-1-ol, 2-bromo-2,3-dihydro, trans-2-bromo-1-indanol, bromoindanol, acmc-20ajgx, pubchem3783
PubChem CID 95444
IUPAC Name 2-bromo-2,3-dihydro-1H-inden-1-ol
SMILES OC1C(Br)CC2=CC=CC=C12

Specifications

Melting Point 130.0°C to 131.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Quantity 250 g
Formula Weight 213.07
Percent Purity 99%
Chemical Name or Material 2-Bromo-1-indanol
EINECSNumber 226-442-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.