Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-naphthol, 95%

Catalog No. AAH6642906
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6642906 5 g
AAH66429MD 250 mg
AAH6642903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6642906 Supplier Thermo Scientific Chemicals Supplier No. H6642906
Only null left
Add to Cart

CAS: 771-15-3 | C10H7BrO | 223.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-15-3
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.07
MDL Number MFCD00274158
InChI Key LOHOSHDZQVKDPS-UHFFFAOYSA-N
Synonym 2-bromo-1-naphthol, 2-bromo-naphthalen-1-ol, 2-bromo-1-naphthalenol, 1-naphthalenol, 2-bromo, naphthalenol, bromo, bromonaphthol, bromnaphthol, pubchem22909, 2-bromanylnaphthalen-1-ol, acmc-209vk0
PubChem CID 150849
IUPAC Name 2-bromonaphthalen-1-ol
SMILES OC1=C2C=CC=CC2=CC=C1Br

Specifications

Melting Point 44°C to 46°C
Color White
Quantity 5 g
Beilstein 2554660
Formula Weight 223.07
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Bromo-1-naphthol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.