Learn More
Cayman Chemical 2-bromo 17β-Estradiol, Purity: ≥98%, CAS: 15833-07-5, 50 mg
SDP
Catalog No. 502893645
Encompass_Preferred
A synthetic steroid and inhibitor of estrogen 2-hydroxylase; inhibits estrogen 2-hydroxylase in rat liver microsomes at 50 and 100 µM; prevents estrogen-induced inhibition of apomorphine-induced climbing behavior in mice at 5 mg/kg
Catalog No. 50-289-3645 Supplier Cayman Chemical Supplier No. 900214550 MG
A synthetic steroid and inhibitor of estrogen 2-hydroxylase; inhibits estrogen 2-hydroxylase in rat liver microsomes at 50 and 100 µM; prevents estrogen-induced inhibition of apomorphine-induced climbing behavior in mice at 5 mg/kg