Learn More
Cayman Chemical 2-bromo 17-EstradIol 5mg
SDP
Catalog No. 502893611
Encompass_Preferred
A synthetic steroid and inhibitor of estrogen 2-hydroxylase; inhibits estrogen 2-hydroxylase in rat liver microsomes at 50 and 100 µM; prevents estrogen-induced inhibition of apomorphine-induced climbing behavior in mice at 5 mg/kg
Catalog No. 50-289-3611 Supplier Cayman Chemical Supplier No. 90021455 MG
A synthetic steroid and inhibitor of estrogen 2-hydroxylase; inhibits estrogen 2-hydroxylase in rat liver microsomes at 50 and 100 µM; prevents estrogen-induced inhibition of apomorphine-induced climbing behavior in mice at 5 mg/kg