Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2',4'-dichloroacetophenone, 95%

Catalog No. AAH5961506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5961506 5 g
AAH5961503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5961506 Supplier Thermo Scientific Chemicals Supplier No. H5961506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2631-72-3 | C8H5BrCl2O | 267.931 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2631-72-3
Molecular Formula C8H5BrCl2O
Molecular Weight (g/mol) 267.931
MDL Number MFCD00053005
InChI Key DASJDMQCPIDJIF-UHFFFAOYSA-N
Synonym 2-bromo-2',4'-dichloroacetophenone, 2-bromo-1-2,4-dichlorophenyl ethanone, 2,4-dichlorophenacyl bromide, 2-bromo-1-2,4-dichlorophenyl ethan-1-one, 2-bromo-1-2,4-dichloro-phenyl-ethanone, ethanone, 2-bromo-1-2,4-dichlorophenyl, 2'-bromo-2,4-dichloro acetophenone, omega-bromo-2,4-dichloro acetophenone, 2',4'-dichlorophenacyl bromide, 2-bromo-2,4-dichloroacetophenone
PubChem CID 75828
IUPAC Name 2-bromo-1-(2,4-dichlorophenyl)ethanone
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)CBr

Specifications

Melting Point 25°C to 29°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN1759
Formula Weight 267.93
Percent Purity 95%
Chemical Name or Material 2-Bromo-2',4'-dichloroacetophenone
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 220-116-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.