Learn More
Medchemexpress LLC 2'-bromo-2'-deoxyuridine | 4753-02-0 | MFCD06658226 | 96.9% | 307.1 g/mol | C9H11BrN2O5 | 10 MG
SDP
Catalog No. 5000282674
Encompass_Preferred
2'-Bromo-2'-deoxyuridine is a brominated nucleoside analog used as a research reagent for DNA synthesis and cell proliferation studies. It acts as a thymidine analog for incorporation into DNA and is supplied as a purified solid with reported HPLC purity of 96.92%.
  • Brominated nucleoside analog for DNA incorporation studies.
  • Used in cell proliferation and DNA replication assays.
  • Supplied as a stable solid for laboratory use.
  • High HPLC purity suitable for research applications.
  • Compatible with standard molecular biology protocols.
Catalog No. 50-002-82674 Supplier Medchemexpress LLC Supplier No. HYW14133810MG
2'-Bromo-2'-deoxyuridine is a brominated nucleoside analog used as a research reagent for DNA synthesis and cell proliferation studies. It acts as a thymidine analog for incorporation into DNA and is supplied as a purified solid with reported HPLC purity of 96.92%.
  • Brominated nucleoside analog for DNA incorporation studies.
  • Used in cell proliferation and DNA replication assays.
  • Supplied as a stable solid for laboratory use.
  • High HPLC purity suitable for research applications.
  • Compatible with standard molecular biology protocols.