Learn More
Medchemexpress LLC 2'-bromo-2'-deoxyuridine | 4753-02-0 | MFCD06658226 | 96.9% | 307.10 | C9H11BrN2O5 | 1 ML
SDP
Catalog No. 5000283320
Encompass_Preferred
2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside analog used in research to label DNA and investigate DNA synthesis, cell proliferation, and apoptosis. It is supplied both as a ready-to-use 10 mM solution (1 mL) and as solid quantities for synthesis and assay use.
  • Brominated deoxyuridine nucleoside analog for DNA labeling and tracking.
  • Supplied as a 10 mM solution in 1 mL and in solid pack sizes for flexibility.
  • High purity (~96.9%) suitable for cellular and biochemical assays.
  • Commonly used to measure DNA synthesis and cell proliferation in vitro.
  • Compatible with standard handling and storage practices for nucleoside reagents.
Catalog No. 50-002-83320 Supplier Medchemexpress LLC Supplier No. HYW14133810MM/1ML
2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside analog used in research to label DNA and investigate DNA synthesis, cell proliferation, and apoptosis. It is supplied both as a ready-to-use 10 mM solution (1 mL) and as solid quantities for synthesis and assay use.
  • Brominated deoxyuridine nucleoside analog for DNA labeling and tracking.
  • Supplied as a 10 mM solution in 1 mL and in solid pack sizes for flexibility.
  • High purity (~96.9%) suitable for cellular and biochemical assays.
  • Commonly used to measure DNA synthesis and cell proliferation in vitro.
  • Compatible with standard handling and storage practices for nucleoside reagents.