Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-bromo-2'-deoxyuridine | 4753-02-0 | MFCD06658226 | 96.9% | 307.10 g/mol | C9H11BrN2O5 | 100 MG
SDP

Catalog No. 5000282673
Encompass_Preferred

2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside supplied for research use. It serves as a nucleoside analog for biochemical and molecular biology studies where modified nucleosides are required.

  • Brominated deoxyuridine nucleoside for incorporation in nucleic acid studies.
  • Molecular formula C9H11BrN2O5; molecular weight 307.10 g/mol.
  • Purity 96.9% by HPLC (certificate of analysis).
  • Available as a powder; storage recommended at -20°C for long-term stability.
  • CAS number 4753-02-0 for chemical identification.
  • Supplied in a 100 mg quantity suitable for lab-scale research.

Catalog No. 50-002-82673 Supplier Medchemexpress LLC Supplier No. HYW141338100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside supplied for research use. It serves as a nucleoside analog for biochemical and molecular biology studies where modified nucleosides are required.

  • Brominated deoxyuridine nucleoside for incorporation in nucleic acid studies.
  • Molecular formula C9H11BrN2O5; molecular weight 307.10 g/mol.
  • Purity 96.9% by HPLC (certificate of analysis).
  • Available as a powder; storage recommended at -20°C for long-term stability.
  • CAS number 4753-02-0 for chemical identification.
  • Supplied in a 100 mg quantity suitable for lab-scale research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.