Learn More
Medchemexpress LLC 2'-bromo-2'-deoxyuridine | 4753-02-0 | MFCD06658226 | 96.9% | 307.10 g·mol⁻¹ | C9H11BrN2O5 | 25 MG
SDP
Catalog No. 5000283440
Encompass_Preferred
2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside analog used in biochemical and cell biology research. Supplied as a solid powder, it is commonly used as an antimetabolite and as a marker to study DNA synthesis, replication, and repair. Certificate of analysis provides purity and storage guidance for research use.
  • Brominated nucleoside analog useful as a DNA synthesis marker.
  • High purity suitable for research applications.
  • White to off-white solid for easy handling and storage.
  • Stable under recommended storage conditions for extended shelf life.
  • Applicable in cell proliferation, DNA repair, and replication studies.
Catalog No. 50-002-83440 Supplier Medchemexpress LLC Supplier No. HYW14133825MG
2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside analog used in biochemical and cell biology research. Supplied as a solid powder, it is commonly used as an antimetabolite and as a marker to study DNA synthesis, replication, and repair. Certificate of analysis provides purity and storage guidance for research use.
  • Brominated nucleoside analog useful as a DNA synthesis marker.
  • High purity suitable for research applications.
  • White to off-white solid for easy handling and storage.
  • Stable under recommended storage conditions for extended shelf life.
  • Applicable in cell proliferation, DNA repair, and replication studies.