Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-bromo-2'-deoxyuridine | 4753-02-0 | MFCD06658226 | 96.9% | 307.10 g/mol | C9H11BrN2O5 | 5 MG
SDP

Catalog No. 5000283323
Encompass_Preferred

2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside analog supplied for research use. It is provided as a white to off-white solid and is commonly used as a thymidine analog for DNA incorporation, cell proliferation assays, and other biochemical studies.

  • White to off-white solid appearance.
  • Molecular formula C9H11BrN2O5; molecular weight 307.10 g/mol.
  • CAS number 4753-02-0.
  • Purity 96.9% (HPLC).
  • Storage: powder at -20°C for long-term stability.
  • Intended for research use only.

Catalog No. 50-002-83323 Supplier Medchemexpress LLC Supplier No. HYW1413385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2'-Bromo-2'-deoxyuridine is a brominated deoxyuridine nucleoside analog supplied for research use. It is provided as a white to off-white solid and is commonly used as a thymidine analog for DNA incorporation, cell proliferation assays, and other biochemical studies.

  • White to off-white solid appearance.
  • Molecular formula C9H11BrN2O5; molecular weight 307.10 g/mol.
  • CAS number 4753-02-0.
  • Purity 96.9% (HPLC).
  • Storage: powder at -20°C for long-term stability.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.