Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 2'-Bromo-2'-deoxyuridine, 98%

Catalog No. AAJ64704ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64704ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64704ME Supplier Thermo Scientific Chemicals Supplier No. J64704ME

missing translation for 'validMainframeMsg'

CAS: 72218-68-9 | C9H11BrN2O5 | 307.10 g/mol

2'-Bromo-2'-deoxyuridine is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell signaling and other processes that induce cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Bromo-2′-deoxyuridine is a thymidine analogue which is incorporated into DNA. 5-BrdU is routinely and extensively used to measure DNA synthesis and to label dividing cells. Consequently 5-BrdU is used to study cell si
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72218-68-9
Molecular Formula C9H11BrN2O5
Molecular Weight (g/mol) 307.10
MDL Number MFCD06658226
InChI Key FEUDNSHXOOLCEY-XVFCMESISA-N
Synonym 2'-bromo-2'-deoxyuridine, 2'-bromo-2'-deoxy-d-uridine, 2'-deoxy-2'-bromo-uridine, 1-2r,3r,4r,5r-3-bromo-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, 1-2r,3r,4r,5r-3-bromo-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, 2'-bromodesoxyuridine, 2'-bromo-2'-deoxyuridin, 2-bromo-2-deoxyuridine, 1-2r,3r,4r,5r-3-bromo-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione
PubChem CID 188257
IUPAC Name 1-[(2R,3R,4R,5R)-3-bromo-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](Br)[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Color White
Quantity 500 mg
Solubility Information Soluble in water.
Formula Weight 307.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Bromo-2'-deoxyuridine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.