Learn More
Medchemexpress LLC 2-Bromo-2'-methoxyacetophenone | 31949-21-0 | MFCD00000196 | 98.4% | 229.07 | 50 G
SDP
Catalog No. 5000452623
Encompass_Preferred
2-Bromo-2'-methoxyacetophenone is an irreversible inhibitor of MurA, inhibiting MurA in *E. coli* with an IC50 of 0.38 μM. It is suitable for antibacterial research aimed at disrupting bacterial cell wall synthesis.
  • Irreversible inhibitor for MurA
  • Inhibits MurA in E. coli with an IC50 of 0.38 μM
  • Can be utilized in antibacterial research
  • Blocks bacterial cell wall synthesis
Catalog No. 50-004-52623 Supplier Medchemexpress LLC Supplier No. HYW01226750G
2-Bromo-2'-methoxyacetophenone is an irreversible inhibitor of MurA, inhibiting MurA in *E. coli* with an IC50 of 0.38 μM. It is suitable for antibacterial research aimed at disrupting bacterial cell wall synthesis.
  • Irreversible inhibitor for MurA
  • Inhibits MurA in E. coli with an IC50 of 0.38 μM
  • Can be utilized in antibacterial research
  • Blocks bacterial cell wall synthesis