Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromo-2-methylbutane

Catalog No. 306878100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-687-8100G 100 g
30-687-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-687-8100G Supplier Sigma Aldrich Supplier No. 306878100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The enthalpies of the solution of 2-bromo-2-methylbutane and 2-bromo-2-methylpropane in mono- and dialcohols were studied.

Specifications

CAS 507-36-8
Density 1.182 g/mL (at 25°C (literature))
Boiling Point 107°C (735 mmHg)
Molecular Formula C5H11Br
Linear Formula CH3CH2CBr(CH3)2
Refractive Index n20/D 1.4423 (literature)
MDL Number MFCD00000127
Quantity 100 g
Synonym tert-Amyl bromide
Molecular Weight (g/mol) 151.04
Percent Purity 95%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.