Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2-methylpropionyl Bromide, 98%

Catalog No. p-3735754
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403090050 5 g Glass bottle
AC403091000 100 g Glass bottle
AC403095000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC403090050 Supplier Thermo Scientific Chemicals Supplier No. 403090050
Only null left
Add to Cart

CAS: 20769-85-1 | C4H6Br2O | 229.9 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20769-85-1
Molecular Formula C4H6Br2O
Molecular Weight (g/mol) 229.9
MDL Number MFCD00000122
InChI Key YOCIJWAHRAJQFT-UHFFFAOYSA-N
Synonym 2-bromo-2-methylpropionyl bromide, 2-bromoisobutyryl bromide, propanoyl bromide, 2-bromo-2-methyl, bibb, 2-bromo-2-methylpropionylbromide, unii-627f9n9dya, 2-bromoisobutyrylbromide, 2-bromo-2-methyl-propionyl bromide, alpha-bromoisobutyryl bromide, 2-bromo-2-methyl-propanoyl bromide
PubChem CID 88685
IUPAC Name 2-bromo-2-methylpropanoyl bromide
SMILES CC(C)(C(=O)Br)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.8600g/mL
Boiling Point 162°C to 164°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5070 to 1.5090
Linear Formula (CH3)2CBrCOBr
Quantity 5 g
Specific Gravity 1.86
Solubility Information Solubility in water: reacts
Formula Weight 229.9
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropionyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
May cause genetic defects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 244-017-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.