Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3,5-dichloropyridine-4-carboxylic acid, 95%, Thermo Scientific™

Catalog No. AAH6373903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6373903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6373903 Supplier Thermo Scientific Chemicals Supplier No. H6373903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 343781-56-6
Molecular Formula C6H2BrCl2NO2
Molecular Weight (g/mol) 270.891
MDL Number MFCD03411637
InChI Key DSKHVZJZZXHWPU-UHFFFAOYSA-N
Synonym 2-bromo-3,5-dichloroisonicotinic acid, pubchem13557, 2-bromo-3,5-dichloro-isonicotinic acid, 4-pyridinecarboxylicacid, 2-bromo-3,5-dichloro
PubChem CID 2762469
IUPAC Name 2-bromo-3,5-dichloropyridine-4-carboxylic acid
SMILES C1=C(C(=C(C(=N1)Br)Cl)C(=O)O)Cl

Specifications

Color White
Quantity 1 g
Formula Weight 270.9
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-3,5-dichloropyridine-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.