Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3,5-dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. B37345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37345G 5 g
B373425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37345G Supplier TCI America Supplier No. B37345G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14482-51-0
Molecular Formula C5H2BrCl2N
Molecular Weight (g/mol) 226.88
MDL Number MFCD00233990
InChI Key QCKPJWDIDCGQRB-UHFFFAOYSA-N
Synonym 3,5-dichloro-2-bromopyridine, 2-bromo-3,5-dichloro-pyridine, pyridine, 2-bromo-3,5-dichloro, pubchem10223, acmc-1bqk7, 2-bromo-3,5-dichloropyridin, ksc494o1r, tpc-py058, 2-bromo-3,5 dichloro pyridine
PubChem CID 817090
IUPAC Name 2-bromo-3,5-dichloropyridine
SMILES ClC1=CN=C(Br)C(Cl)=C1

Specifications

Melting Point 43°C
Quantity 5 g
Formula Weight 226.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3,5-dichloropyridine
EINECSNumber (5)-5257
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.