Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-butylthiophene 98.0+%, TCI America™
SDP

Catalog No. B45115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B45115G Supplier TCI America Supplier No. B45115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 145543-82-4
Molecular Formula C8H11BrS
Molecular Weight (g/mol) 219.14
MDL Number MFCD16619253
InChI Key KNBLITSBUDZSJW-UHFFFAOYSA-N
PubChem CID 15354891
IUPAC Name 2-bromo-3-butylthiophene
SMILES CCCCC1=C(SC=C1)Br

Specifications

Color Yellow
Boiling Point 59°C
Quantity 5 g
Formula Weight 219.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-butylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.