Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-dodecylthiophene 97.0+%, TCI America™
SDP

Catalog No. B36915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36915G 5 g
B36911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36915G Supplier TCI America Supplier No. B36915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 139100-06-4
Molecular Formula C16H27BrS
Molecular Weight (g/mol) 331.36
MDL Number MFCD10000891
InChI Key IMILVTHOOISGRW-UHFFFAOYSA-N
Synonym 2-Bromo-3-laurylthiophene
PubChem CID 10426816
IUPAC Name 2-bromo-3-dodecylthiophene
SMILES CCCCCCCCCCCCC1=C(Br)SC=C1

Specifications

Boiling Point 165°C
Quantity 5 g
Formula Weight 331.36
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-dodecylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.