Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromo-3-methoxyisonicotinaldehyde

Catalog No. ADE0001761G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0001761G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0001761G Supplier Sigma Aldrich Supplier No. ADE0001761G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,2,3-triazines have been shown to be reactive substrates in inverse electron demand diels-alder strategies
  • Recent examples by the-ßboger research group-ßhave utilized this reactive motif in the construction of highly functionalized n-containing heterocycles.total syntheses of (-)-pyrimidoblamic acid and p-3a

Specifications

CAS 191418-78-7
Molecular Formula C7H6BrNO2
Linear Formula C7H6BrNO2
MDL Number MFCD06410674
Quantity 100 mg
Molecular Weight (g/mol) 216.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.