Learn More
Medchemexpress LLC 2-Bromo-3-methylpyridine | 3430-17-9 | 172.03 | 1 KG
SDP
Catalog No. 5000446479
Encompass_Preferred
2-Bromo-3-methylpyridine is a heterocyclic intermediate belonging to the halomethylpyridine family. It can be utilized in the synthesis of platinum(IV) diazide complexes.
  • Heterocyclic intermediate
  • Belongs to the halomethylpyridine family
  • Can be utilized in the synthesis of platinum(IV) diazide complexes
Catalog No. 50-004-46479 Supplier Medchemexpress LLC Supplier No. HYW0077121KG
2-Bromo-3-methylpyridine is a heterocyclic intermediate belonging to the halomethylpyridine family. It can be utilized in the synthesis of platinum(IV) diazide complexes.
  • Heterocyclic intermediate
  • Belongs to the halomethylpyridine family
  • Can be utilized in the synthesis of platinum(IV) diazide complexes