Learn More
Medchemexpress LLC 2-Bromo-3-methylpyridine | 3430-17-9 | 172.03 | 100 G
SDP
Catalog No. 5000448700
Encompass_Preferred
2-Bromo-3-methylpyridine is a heterocyclic intermediate of the halomethylpyridine family. It can be used in the synthesis of platinum(IV) diazide complexes.
  • Heterocyclic intermediate
  • Used in synthesis of platinum(IV) diazide complexes
Catalog No. 50-004-48700 Supplier Medchemexpress LLC Supplier No. HYW007712100G
2-Bromo-3-methylpyridine is a heterocyclic intermediate of the halomethylpyridine family. It can be used in the synthesis of platinum(IV) diazide complexes.
  • Heterocyclic intermediate
  • Used in synthesis of platinum(IV) diazide complexes