Learn More
Medchemexpress LLC 2-bromo-3-methylpyridine | 3430-17-9 | 172.03 | 50 G
SDP
Catalog No. 5000446348
Encompass_Preferred
2-Bromo-3-methylpyridine (2-Bromo-3-picoline) is a heterocyclic intermediate of the halomethylpyridine family. It can be used in the synthesis of platinum(IV) diazide complexes.
  • Heterocyclic intermediate
  • Used in the synthesis of platinum(IV) diazide complexes
  • For research use only
  • Appears as a liquid
  • Colorless to light yellow in appearance
Catalog No. 50-004-46348 Supplier Medchemexpress LLC Supplier No. HYW00771250G
2-Bromo-3-methylpyridine (2-Bromo-3-picoline) is a heterocyclic intermediate of the halomethylpyridine family. It can be used in the synthesis of platinum(IV) diazide complexes.
  • Heterocyclic intermediate
  • Used in the synthesis of platinum(IV) diazide complexes
  • For research use only
  • Appears as a liquid
  • Colorless to light yellow in appearance