Learn More
Medchemexpress LLC 2-Bromo-3-methylpyridine | 3430-17-9 | MFCD00239380 | 172.03 | 500 G
SDP
Catalog No. 5000448701
Encompass_Preferred
2-Bromo-3-methylpyridine (2-Bromo-3-picoline) is a heterocyclic intermediate of the halomethylpyridine family. It can be used in the synthesis of platinum(IV) diazide complexes, and serves as a versatile building block in the synthesis of various pharmaceuticals and agrochemicals.
  • Heterocyclic intermediate
  • Used in the synthesis of platinum(IV) diazide complexes
  • Versatile building block for pharmaceuticals and agrochemicals
  • Storage conditions: Keep container tightly closed, store in a cool and dark place, away from heat, moisture, incompatible chemicals, excessive heat, and light. Store under inert gas, as it is air sensitive.
Catalog No. 50-004-48701 Supplier Medchemexpress LLC Supplier No. HYW007712500G
2-Bromo-3-methylpyridine (2-Bromo-3-picoline) is a heterocyclic intermediate of the halomethylpyridine family. It can be used in the synthesis of platinum(IV) diazide complexes, and serves as a versatile building block in the synthesis of various pharmaceuticals and agrochemicals.
  • Heterocyclic intermediate
  • Used in the synthesis of platinum(IV) diazide complexes
  • Versatile building block for pharmaceuticals and agrochemicals
  • Storage conditions: Keep container tightly closed, store in a cool and dark place, away from heat, moisture, incompatible chemicals, excessive heat, and light. Store under inert gas, as it is air sensitive.