Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-n-octylthiophene 97.0+%, TCI America™
SDP

Catalog No. B37335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37335G Supplier TCI America Supplier No. B37335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 145543-83-5
Molecular Formula C12H19BrS
Molecular Weight (g/mol) 275.248
MDL Number MFCD13193238
InChI Key ISONQKSIWXLJOQ-UHFFFAOYSA-N
PubChem CID 10754854
IUPAC Name 2-bromo-3-octylthiophene
SMILES CCCCCCCCC1=C(SC=C1)Br

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
Formula Weight 275.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-n-octylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.