Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-pyridinemethanol 98.0+%, TCI America™
SDP

Catalog No. B26741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26741G Supplier TCI America Supplier No. B26741G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 131747-54-1
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
InChI Key QIQZUABMLGJKJP-UHFFFAOYSA-N
Synonym 2-bromopyridin-3-yl methanol, 2-bromo-3-pyridinemethanol, 2-bromo-3-hydroxymethyl pyridine, 2-bromo-3-pyridyl methanol, 3-pyridinemethanol, 2-bromo, 2-bromo-3-hydroxymethylpyridine, 2-bromopyridine-3-methanol, 2-bromonicotinyl alcohol, acmc-1c5gr, 2-bromo-pyridin-3-yl-methanol
PubChem CID 14761460
IUPAC Name (2-bromopyridin-3-yl)methanol
SMILES C1=CC(=C(N=C1)Br)CO

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-pyridinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.