Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-tetradecylthiophene 95.0+%, TCI America™
SDP

Catalog No. B54085G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54085G 5 g
B54081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54085G Supplier TCI America Supplier No. B54085G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 500199-09-7
Molecular Formula C18H31BrS
Molecular Weight (g/mol) 359.41
InChI Key GOUPSYVDGUEWMF-UHFFFAOYSA-N
PubChem CID 67293590
IUPAC Name 2-bromo-3-tetradecylthiophene
SMILES CCCCCCCCCCCCCCC1=C(SC=C1)Br

Specifications

Quantity 5 g
Formula Weight 359.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-tetradecylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.