Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4,5-dimethoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B20115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20115G 5 g
B201125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20115G Supplier TCI America Supplier No. B20115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5392-10-9
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.07
MDL Number MFCD00003301
InChI Key UQQROBHFUDBOOK-UHFFFAOYSA-N
Synonym 6-bromoveratraldehyde, 6-bromo-3,4-dimethoxybenzaldehyde, benzaldehyde, 2-bromo-4,5-dimethoxy, 6-bromo veratraldehyde, 2-bromo-4,5-dimethoxybenzalehyde, 4,5-dimethoxy-2-bromobenzaldehyde, 6-bromoveratral, pubchem8209, veratraldehyde, 6-bromo, acmc-2097hh
PubChem CID 79351
IUPAC Name 2-bromo-4,5-dimethoxybenzaldehyde
SMILES COC1=C(C=C(C(=C1)C=O)Br)OC

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 245.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4,5-dimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.