Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluoro-5-nitrophenol, 97%, Thermo Scientific™

Catalog No. AAH6680103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6680103 1 g
AAH6680106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6680103 Supplier Thermo Scientific Chemicals Supplier No. H6680103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84478-87-5
Molecular Formula C6H3BrFNO3
Molecular Weight (g/mol) 236.00
MDL Number MFCD08460104
InChI Key NVNFKCCUJKPLLT-UHFFFAOYSA-N
Synonym phenol, 2-bromo-4-fluoro-5-nitro, 2-bromo-4-fluoro-5-nitro-phenol, 4-bromo-2-fluoro-5-hydroxynitrobenzene, pubchem2837, ksc495s7j, 2-bromo-4-fluro-5-nitrophenol
PubChem CID 13268179
IUPAC Name 2-bromo-4-fluoro-5-nitrophenol
SMILES OC1=C(Br)C=C(F)C(=C1)[N+]([O-])=O

Specifications

Melting Point 124°C to 127°C
Color Brown
Quantity 1 g
UN Number UN2811
Beilstein 15697238
Formula Weight 236
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Bromo-4-fluoro-5-nitrophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.