Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluorobenzonitrile, 98%, Thermo Scientific Chemicals

Catalog No. AAL2011506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2011506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2011506 Supplier Thermo Scientific Chemicals Supplier No. L2011506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 36282-26-5 | Molecular Formula: C7H3BrFN | Molecular Weight: 200.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36282-26-5
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00672924
InChI Key MNNDREXLRLDWEY-UHFFFAOYSA-N
Synonym 4-fluoro-2-bromobenzonitrile, 2-bromo-4-fluoro-benzonitrile, 2-bromo-4-fluorobenzenecarbonitrile, 2-cyano-5-fluorobromobenzene, benzonitrile, 2-bromo-4-fluoro, pubchem3217, acmc-1ajip, ksc490q6h, 2-bromo-4-fluoro benzonitrile, 2-bromo-4-fluorobenzonitrile
PubChem CID 118939
IUPAC Name 2-bromo-4-fluorobenzonitrile
SMILES FC1=CC=C(C#N)C(Br)=C1

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
UN Number UN3439
Beilstein 1940597
Formula Weight 200.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 252-949-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.