Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluorobenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AAH6185014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6185014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6185014 Supplier Thermo Scientific Chemicals Supplier No. H6185014

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromo-4-fluorobenzyl alcohol, is used as an alcohol, medicine intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 229027-89-8
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.03
MDL Number MFCD00672925
InChI Key XGAMLBPEVCLQEJ-UHFFFAOYSA-N
PubChem CID 2773347
IUPAC Name (2-bromo-4-fluorophenyl)methanol
SMILES OCC1=C(Br)C=C(F)C=C1

Specifications

Melting Point 68°C to 72°C
Color White
Quantity 25 g
Synonym 2-bromo-4-fluorophenyl methanol, 2-bromo-4-fluorobenzyl alcohol, benzenemethanol, 2-bromo-4-fluoro, 2-bromo-4-fluorophenyl methan-1-ol, 2-bromo-4-fluorobenzenemethanol, benzenemethanol,2-bromo-4-fluoro, pubchem3222, acmc-1cqar, ksc550i1h, rarechem al bd 1088
Solubility Information Soluble in water.
Formula Weight 205.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-4-fluorobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.