Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Bromo-4-fluorophenol | 496-69-5 | 191.00 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW003971

Encompass_Preferred

2-Bromo-4-fluorophenol is a biochemical reagent intended for life science related research. It is an organic compound with the molecular formula C6H4BrFO.

  • Biochemical reagent
  • Suitable for life science research
  • Molecular formula: C6H4BrFO
  • Molecular weight: 191.00
  • CAS number: 496-69-5
  • High purity
  • For research use only

Catalog No. 50-004-49413


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.